missing translation for 'onlineSavingsMsg'
Learn More

SIRP beta 1/CD172b Antibody (308908), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18580266 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18580266 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18580266 Supplier Novus Biologicals Supplier No. FAB20961X

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SIRP beta 1/CD172b Monoclonal antibody specifically detects SIRP beta 1/CD172b in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRP beta 1/CD172b
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 308908
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD172 antigen-like family member B, CD172b, CD172b antigen, DKFZp686A05192, FLJ26614, signal-regulatory protein beta 1, signal-regulatory protein beta-1, SIRP-BETA-1, SIRP-beta-1 isoform 3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human SIRP beta 1/CD172b, Gly26-Ala369 (Arg53His and Ala363Pro), Accession # O00241.4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10326
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.