missing translation for 'onlineSavingsMsg'
Learn More

TREM1 Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™

Product Code. 15296845
Change view
Click to view available options
Quantity:
5 μg
Unit Size:
5µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15296845 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15296845 Supplier Invitrogen Supplier No. 10511H03H5

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 200) of human TREM1 (NP_061113.1) precursor fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Triggering Receptor Expressed on Myeloid Cells 1 (TREM1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
Gene ID (Entrez) 54210
Molecular Weight (g/mol) 48.3kDa
Purification Method PAGE
Quantity 5 μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination C-Terminal
Structural Form Truncated/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias TREM-1, TREM1
Common Name Triggering receptor expressed on myeloid cells 1
Gene Symbol TREM1
Protein Tag IgG1 Fc Tag
Expression System Mammalian
Protein Family Other Proteins
Form Lyophilized
Protein Subtype Other Proteins
Purity or Quality Grade ≥90% by SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.