Learn More
Applied Biosystems™ Single-Tube TaqMan™ SNP Genotyping Assays
Product Code. 10176954 Shop All Applied Biosystems Products
Change view
Click to view available options
L (2400 reactions), made to order
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10176954 L (2400 reactions), made to order
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10176954 Supplier Applied Biosystems™ Supplier No. 4351380
  • ONLINE EXCLUSIVE PRICE
    1566.00 EUR
    1017.90 EUR / Pack of 1
    Save 548.10 EUR
    35% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Includes
Two unlabeled PCR primers: (forward and reverse primers at 900nM final concentration); One Applied Biosystems VIC™ dye: MGB labeled probe detects the Allele 1 sequence (probes at 200nM final concentration); One Applied Biosystems 6FAM™ dye: MGB labeled probe detects the Allele 2 sequence (probes at 200nM final concentration)
5' nuclease assay to discriminate between two alleles of a specific SNP for use in genotyping studies
  • Large selection of SNP assays advances validation and screening studies for disease association and population stratification applications
  • Easy-to-use, single-tube format
  • Robust assay design provides highly accurate, reproducible and reliable results
  • Simple workflow enables quick results
  • Large selection of SNP assays advances validation and screening studies for disease association and population stratification applications
  • Easy-to-use, single-tube format
  • Robust assay design provides highly accurate, reproducible and reliable results
  • Simple workflow enables quick results
TRUSTED_SUSTAINABILITY
Specifications
Concentration 80X
Content And Storage Two unlabeled PCR primers: (forward and reverse primers at 900nM final concentration); One Applied Biosystems VIC™ dye: MGB labeled probe detects the Allele 1 sequence (probes at 200nM final concentration); One Applied Biosystems 6FAM™ dye: MGB labeled probe detects the Allele 2 sequence (probes at 200nM final concentration). Store at -15° to -25°C
Description Single-Tube TaqMan SNP Genotyping Assays
Quantity L (2400 reactions), made to order
Species Non-human
For Use With (Application) Genotyping
No. of Reactions 2400 reactions
Sample Type Mouse