Learn More
- Home
- Products
- Furniture
- Desks and Tables
- Laboratory Benches and Tables
- Köttermann Single Bench
Köttermann Single Bench
Product Code. 17392219
Change view
Click to view available options
Dimensions (D x W x H):
600 x 900 x 750 mm
600 x 900 x 900 mm
750 x 900 x 750 mm
750 x 900 x 900 mm
600 x 1200 x 750 mm
600 x 1200 x 900 mm
600 x 1500 x 750 mm
600 x 1500 x 900 mm
750 x 1200 x 750 mm
750 x 1200 x 900 mm
750 x 1500 x 750 mm
750 x 1500 x 900 mm
Top Style:
Melamine TopResist Worktop
Trespa TopLab PLUS Worktop
Unit Size:
Each
24 product options available for selection
| Product Code. | Dimensions (D x W x H) | Top Style | unitSize |
|---|---|---|---|
| 17392219 | 750 x 900 x 900 mm | Trespa TopLab PLUS Worktop | Each |
| 17372189 | 600 x 900 x 750 mm | Melamine TopResist Worktop | Each |
| 17302219 | 600 x 900 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17302199 | 600 x 900 x 900 mm | Melamine TopResist Worktop | Each |
| 17332219 | 600 x 900 x 900 mm | Trespa TopLab PLUS Worktop | Each |
| 17382189 | 600 x 1200 x 750 mm | Melamine TopResist Worktop | Each |
| 17312219 | 600 x 1200 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17312199 | 600 x 1200 x 900 mm | Melamine TopResist Worktop | Each |
| 17342219 | 600 x 1200 x 900 mm | Trespa TopLab PLUS Worktop | Each |
| 17392189 | 600 x 1500 x 750 mm | Melamine TopResist Worktop | Each |
| 17322219 | 600 x 1500 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17322199 | 600 x 1500 x 900 mm | Melamine TopResist Worktop | Each |
| 17352219 | 600 x 1500 x 900 mm | Trespa TopLab PLUS Worktop | Each |
| 17332199 | 750 x 900 x 750 mm | Melamine TopResist Worktop | Each |
| 17362219 | 750 x 900 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17362199 | 750 x 900 x 900 mm | Melamine TopResist Worktop | Each |
| 17342199 | 750 x 1200 x 750 mm | Melamine TopResist Worktop | Each |
| 17372219 | 750 x 1200 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17372199 | 750 x 1200 x 900 mm | Melamine TopResist Worktop | Each |
| 17302229 | 750 x 1200 x 900 mm | Trespa TopLab PLUS Worktop | Each |
| 17352199 | 750 x 1500 x 750 mm | Melamine TopResist Worktop | Each |
| 17382219 | 750 x 1500 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17382199 | 750 x 1500 x 900 mm | Melamine TopResist Worktop | Each |
| 17312229 | 750 x 1500 x 900 mm | Trespa TopLab PLUS Worktop | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
This item is not returnable.
View return policy
Product Code.
17392219
Supplier
Köttermann
Supplier No.
50500162
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable.
View return policy
Description
- Available for sitting and standing work height
- Stationary
Specifications
Specifications
| Dimensions (D x W x H) | 750 x 900 x 900 mm |
| No. of Shelves | 1 |
| Top Style | Trespa TopLab PLUS Worktop |
Product Title
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?