missing translation for 'onlineSavingsMsg'
Learn More

Simvastatin EP Impurity B, Mikromol™

Product Code. 30050347
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050347 25 mg 25mg
30051592 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050347 Supplier Mikromol Supplier No. MM0158.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Simvastatin EP Impurity B
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetyl Simvastatin
CAS 145576-25-6
Analyte or Component Names Acetyl Simvastatin
Shipping Condition Room Temperature
Molecular Formula C27 H40 O6
Certifications/Compliance ISO 17025
Synonym (1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-(Acetyloxy)-6-oxotetrahydro-2H-pyran-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2,2-Dimethylbutanoate (Simvastatin Acetate Ester), Simvastatin Imp. B (EP), Acetyl Simvastatin, (1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-(Acetyloxy)-6-oxotetrahydro-2H-pyran-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2,2-Dimethylbutanoate, Simvastatin Acetate Ester, Simvastatin Impurity B
InChI Formula InChI=1S/C27H40O6/c1-7-27(5,6)26(30)33-23-13-16(2)12-19-9-8-17(3)22(25(19)23)11-10-20-14-21(31-18(4)28)15-24(29)32-20/h8-9,12,16-17,20-23,25H,7,10-11,13-15H2,1-6H3/t16-,17-,20+,21+,22-,23-,25-/m0/s1
SMILES CCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@H](CC(=O)O3)OC(=O)C)[C@@H]12
IUPAC Name [(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-acetyloxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] 2,2-dimethylbutanoate
Molecular Weight (g/mol) 460.6
Quantity 25 mg
Formula Weight 460.2825 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.