Learn More
Stomacher™ Simplettes
Product Code. 17774399
Change view
Click to view available options
Length (Metric):
177 mm
250 mm
Unit Size:
Pack of 1000
Pack of 2000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
17774399 250 mm Pack of 1000
17784399 177 mm Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17774399 Supplier Stomacher™ Supplier No. BA6052
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
The Simplettes™ provide an accurate and economical alternative to pipette tips. They are designed specifically to pipette samples from the bottom of the Stomacher™ bag without risk of cross contamination.
  • Single use pipetting straws
  • Capable of pipetting volumes between 0.1 and 2 mL
  • Irradiated sterile
  • Available in various pack sizes
Specifications
Product Type Tip
Diameter (Metric) 4.1 to 4.2 mm (Internal diameter)
Height (Metric) 177 mm
Length (Metric) 250 mm