missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, TRC

Product Code. 30008468
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008468 1 g 1g
30006466 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008468 Supplier Toronto Research Chemicals Supplier No. TRCS4650151G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula Ag . N O3
Molecular Weight (g/mol) 169.87
Synonym Silver nitrate, Nitric acid silver(1+) salt (8CI,9CI), Silver nitrate (7CI), Nitric acid silver(I) salt, Nitric acid, silver(1+) salt, Silver (I) Nitrate, Silver Mononitrate, Silver Nitrate (AgNO3), Silver(1+) Nitrate
IUPAC Name silver;nitrate
SMILES [Ag+].[O-][N+](=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/Ag.NO3/c;2-1(3)4/q+1;-1
Formula Weight 168.8929
Chemical Name or Material Silver nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.