Learn More
Silver Nitrate Solution, 0.1M, Chem-Lab
Product Code. 10080452
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. 10080452
Brand: Chem-Lab CL05.2603.1000
Designed for laboratory use in titrations. Silver Nitrate Solution, 0.1M, Chem-Lab has a concentration of 16.987g AgNO3 per 1L H2O. It is used in chloride determination, as a precursor to other silver compounds and has disinfectant properties.
Specifications
CAS 7761-88-8, 7732-18-5
Quantity 1 L
Molecular Formula AgNO3
MDL Number MFCD00003414
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES [Ag+].[O-][N+]([O-])=O
IUPAC Name silver(1+) nitrate
Molecular Weight (g/mol) 169.87
PubChem CID 24470
ChEBI CHEBI:32130
Formula Weight 169.87g/mol
Concentration or Composition (by Analyte or Components) 16.987g AgNO3 per 1L H2O
Density 1.01g/mL
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Silver nitrate 0.1 mol/l
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P391-Collect spillage.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : silver nitrate