missing translation for 'onlineSavingsMsg'
Learn More

Silver Diethyldithiocarbamate, For Analysis, ACS, Reag. Ph Eur Grades, EMSURE™, MilliporeSigma™

Product Code. 15884080
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15884080 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15884080 Supplier Supelco Supplier No. 1015150005
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 178°C to 183°C
CAS 1470-61-7
Color Yellow
Physical Form Solid
Quantity 5 g
Molecular Formula C5H10AgNS2
Linear Formula (C2H5)2NCS2Ag
Synonym silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate
InChI Key NSVHDIYWJVLAGH-UHFFFAOYSA-M
SMILES CCN(CC)C(=S)[S-].[Ag+]
IUPAC Name silver;N,N-diethylcarbamodithioate
Molecular Weight (g/mol) 256.13
PubChem CID 3034078
Formula Weight 256.14. (molar mass)
Grade ACS/Reagent Ph. Eur.
Identification Passes Test
Odor Characteristic odor
Packaging Glass bottle
Appearance Yellow solid
Chemical Name or Material Silver diethyldithiocarbamate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.