missing translation for 'onlineSavingsMsg'
Learn More

Silver chromate, 99% min

Product Code. p-7050964
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11389237 25 g 25g
11379237 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389237 Supplier Thermo Scientific Alfa Aesar Supplier No. 040107.14
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7784-01-2 | Ag2CrO4 | 331.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-01-2
Molecular Formula Ag2CrO4
Molecular Weight (g/mol) 331.73
MDL Number MFCD00003402
InChI Key OJKANDGLELGDHV-UHFFFAOYSA-N
Synonym silver chromate, unii-zj5092lwu9, silver chromate vi, silberchromat, silver i chromate, 2ag.cro4, chromic acid h2cro4 , disilver 1+ salt, chromic acid h2cro4 , silver 1+ salt 1:2, chromic acid, disilver i salt, disilver dioxido dioxo chromium
PubChem CID 62666
IUPAC Name disilver;dioxido(dioxo)chromium
SMILES [O-][Cr](=O)(=O)[O-].[Ag+].[Ag+]

Specifications

Physical Form Powder
Quantity 25 g
Odor Odorless
Sensitivity Light sensitive
Density 5.63 g/mL
UN Number UN1479
Merck Index 14,8510
Solubility Information Slightly soluble in water; Soluble in HNO3 and ammonia
Formula Weight 331.73
Percent Purity ≤99%
Chemical Name or Material Silver chromate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.