missing translation for 'onlineSavingsMsg'
Learn More

Silver chloride 99+, Supelco™

Product Code. 32239106
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32239106 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32239106 Supplier Supelco Supplier No. 1192031000
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

For analysis 99+ EMSURE™

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silver chloride 99+
Melting Point 455°C
CAS 7783-90-6
Quantity 1 kg
Density 5.56 g/cm³ at 20°C
Synonym Silver chloride 99+, Silver chloride 99+
Molecular Weight (g/mol) 143.32
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.