missing translation for 'onlineSavingsMsg'
Learn More

Silver arsenate, 99.99% (metals basis), Thermo Scientific™

Product Code. p-7059231
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11319748

Brand: Thermo Scientific Alfa Aesar 044228.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13510-44-6
Molecular Formula Ag3AsO4
Molecular Weight (g/mol) 462.522
MDL Number MFCD00046163
InChI Key IMGNYAPMSDUASV-UHFFFAOYSA-K
Synonym silver arsenate, unii-798q5px67i, trisilver arsenate, silberarsenat, trisilver arsorate, 3ag.aso4, arsenic acid h3aso4 , trisilver 1+ salt, arsenic acid h3aso4 , silver 1+ salt 1:3, the arsenic acid h3aso4 ,silver 1+ salt 1:3
PubChem CID 166835
IUPAC Name trisilver;trioxido(oxo)-$l^{5}-arsane
SMILES [O-][As](=O)([O-])[O-].[Ag+].[Ag+].[Ag+]

Specifications

Color Red
Physical Form Lump/Powder
Quantity 5 g
UN Number UN1557
Formula Weight 462.53
Percent Purity 99.99%
Sensitivity Light sensitive
Assay (metals basis)
Chemical Name or Material Silver arsenate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.