accessibility menu, dialog, popup

UserName

CAS RN 13510-44-6

AgAgAgOAsOOO

CAS RN 13510-44-6

IUPAC Name: trisilver(1+) arsorate
Synonyms: silver arsenate
Molecular Weight (g/mol): 462.52
Molecular Formula: Ag3AsO4
InChi Key: IMGNYAPMSDUASV-UHFFFAOYSA-K
SMILES: [Ag+].[Ag+].[Ag+].[O-][As]([O-])([O-])=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results