Learn More
Uvex Silv-Air Classic 3220 FFP2 Mask
Product Code. 17905466 Shop All Uvex Products
Change view
Click to view available options
Filter Type:
FFP2
Unit Size:
Pack of 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filter Type Unit Size
17905466 FFP2 Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17905466 Supplier Uvex Supplier No. 8763220
  • ONLINE EXCLUSIVE PRICE
    41.00 EUR
    40.27 EUR / Pack of 15
    Save 0.73 EUR
    1% Off
Estimated Shipment: 08-12-2026
The uvex silv-Air c product range consists of both classic folding and cup-style respirators that fit a wide variety of facial shapes.
  • With headband loops and integrated sealing lips in all protection classes, it is really easy to fit the masks correctly.
  • The masks can also be worn comfortably for a longer period of time without becoming bothersome.
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type P - Oil-Proof
Color Gray
Design Type Flat-fold
Disposable Reusable
Filter Type FFP2
Material Polypropylene, Polyester
Product Type Healthcare Particulate Respirator and Surgical Mask
Size Medium
With Exhalation Valve Yes
Latex Free Yes