Learn More
Buerkle™ Silo Drill
Product Code. 10607055 Shop All Bürkle Products
Change view
Click to view available options
Material:
Aluminum
Stainless Steel (AISI 304)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10607055 Stainless Steel (AISI 304) Each
10535932 Aluminum Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10607055 Supplier Buerkle™ Supplier No. 53012000
Estimated Shipment: 01-12-2026
If bulk goods are transported over long distances (in trucks, railway goods wagons), separation may occur.Taking the sample just from the surface therefore leads to the wrong result.Representative samples from all layers of the silo can be taken with the SiloDrill.
Representative samples from all layers of a storage or materials silo can be taken with the SiloDrill. The SiloDrill fits through most standard openings with a diameter of 90 mm.
  • Standard length 150 cm, extendable in increments of 100 cm, max. 350 cm
  • Sprung inner tube prevents jamming of coarse grains
  • Scraper edge for cohesive substances
  • Sturdy, removable T-handle
  • Drill tip with drilling blades for easy boring
  • Easy removal of the sample chamber, simple cleaning
TRUSTED_SUSTAINABILITY
Specifications
Product Type Drill Sampler
Material Stainless Steel (AISI 304)
Autoclavable Autoclavable
Capacity (English) 13.53 oz.
Capacity (Metric) 400 mL
Diameter (Metric) Outer Top 40 mm
Length (Metric) 150 cm