Learn More
Lenz Laborglasinstrumente™ Silicone Sealing Ring
Product Code. 12393379
Change view
Click to view available options
Diameter (Metric) Outside:
12 mm
16 mm
22 mm
42 mm
Bore Size:
6 mm
8 mm
12 mm
26 mm
Unit Size:
Each
Pack of 10
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside Bore Size Unit Size
12393379 16 mm 8 mm Pack of 10
10294521 22 mm 12 mm Pack of 10
10528801 42 mm 26 mm Pack of 10
12383379 16 mm 6 mm Pack of 10
11556884 16 mm 8 mm Each
10345071 22 mm 8 mm Pack of 10
12373379 12 mm 6 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12393379 Supplier Lenz Laborglasinstrumente™ Supplier No. 01331808
  • ONLINE EXCLUSIVE PRICE
    38.00 EUR
    30.45 EUR / Pack of 10
    Save 7.55 EUR
    19% Off
Estimated Shipment: 02-11-2026
For protection against leaks in the laboratory. Lenz Laborglasinstrumente™ Silicone Sealing Rings are designed for use with screw caps.
  • Material: silicone rubber (VMQ)
  • With bonded PTFE face
Specifications
Bore Size 8 mm
For Use With (Application) Tubing Diameter: 7.5 to 9.0 mm
Quantity 10/Pk.
Diameter (Metric) Outside 16 mm
Material Silicone Rubber
Type Seal Ring