Learn More
Fisherbrand™ Silicone Pipette Adapter with Safety Valve
Product Code. 15830043 Shop All Fisherbrand Products
Change view
Click to view available options
Adapter with safety valve
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15830043 Adapter with safety valve
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15830043
  • ONLINE EXCLUSIVE PRICE
    8.65 EUR
    6.65 EUR / Pack of 1
    Save 2.00 EUR
    23% Off
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
For use with the Accumax Pipette Controller.
  • Manufactured from silicone
  • With safety valve to provide protection against fluid penetration
  • Aids in fast and accurate pipetting
Specifications
Type Adapter
For Use With (Application) Accurate pipetting
For Use With (Equipment) Accumax Pipette Controller
Includes Safety valve