Learn More
Saint Gobain Life Sciences Silicon Piston Ring
Product Code. 15869272
Change view
Click to view available options
For Use With (Equipment):
2 to 6 L Flask
50 to 500 mL Flask
500 to 2000 mL Flask
Unit Size:
Each
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15869272 50 to 500 mL Flask Each
15859312 500 to 2000 mL Flask Each
11704608 2 to 6 L Flask Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15869272 Supplier Saint Gobain Life Sciences Supplier No. 161040
  • ONLINE EXCLUSIVE PRICE
    25.62 EUR
    21.12 EUR / Pack of 1
    Save 4.50 EUR
    17% Off
View all promotions
Estimated Shipment: 08-12-2026
Use this piston ring for a variety of applications. Silicon Piston Ring is available in a full complement of solid and split joint configurations.
  • Sturdy rubber Silicone piston rings for flasks
  • For general lab applications
TRUSTED_SUSTAINABILITY
Specifications
Material Silicon
For Use With (Equipment) 50 to 500 mL Flask
No. per Pack 1 Ea.
Quantity 1 Ea.
Product Type Piston Ring