Learn More
Saint Gobain Life Sciences Silicon Piston Ring
Product Code. 15859312
Change view
Click to view available options
For Use With (Equipment):
2 to 6 L Flask
50 to 500 mL Flask
500 to 2000 mL Flask
Unit Size:
Each
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15859312 500 to 2000 mL Flask Each
15869272 50 to 500 mL Flask Each
11704608 2 to 6 L Flask Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15859312 Supplier Saint Gobain Life Sciences Supplier No. 161042
  • ONLINE EXCLUSIVE PRICE
    36.00 EUR
    27.96 EUR / Pack of 1
    Save 8.04 EUR
    22% Off
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
Use this piston ring for a variety of applications. Silicon Piston Ring is available in a full complement of solid and split joint configurations.
  • Sturdy rubber Silicone piston rings for flasks
  • For general lab applications
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Material Silicon
For Use With (Equipment) 500 to 2000 mL Flask
No. per Pack 1 Ea.
Quantity 1 Ea.
Product Type Piston Ring