missing translation for 'onlineSavingsMsg'
Learn More

Silicon Dioxide, TRC

Product Code. 30012246
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012246 5 g 5g
30012020 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012246 Supplier Toronto Research Chemicals Supplier No. TRCS4525035G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2 Si
Molecular Weight (g/mol) 60.0843
Synonym Silica, Silica colloidal anhydrous, Silicium dioxide
IUPAC Name Silicon dioxide
SMILES O=[Si]=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/O2Si/c1-3-2
Formula Weight 59.9668
Chemical Name or Material Silicon dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.