missing translation for 'onlineSavingsMsg'
Learn More

Silicic Acid, Supelco™

Product Code. 15866870
Change view
Click to view available options
Quantity:
1 kg
10 kg
Unit Size:
10kg
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15866870 10 kg 10kg
15823930 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15866870 Supplier Supelco Supplier No. 1006579010
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Precipitated extra pure light

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10279-57-9

Specifications

Melting Point 1710°C
Density 2 to 6 g/cm3 at 20°C
Quantity 10 kg
MDL Number MFCD00011232
Synonym Silicic acid, Silicic acid
Molecular Weight (g/mol) 60.08 (anhydrous basis)
Grade Ungraded
Chemical Name or Material Silicic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.