missing translation for 'onlineSavingsMsg'
Learn More

Silicic Acid, Supelco™

Product Code. 15856870
Change view
Click to view available options
Quantity:
20 kg
Unit Size:
20kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15856870 20 kg 20kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15856870 Supplier Supelco Supplier No. 1006569020
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Precipitated extra pure heavy

TRUSTED_SUSTAINABILITY

Specifications

CAS 10279-57-9
Melting Point 1710°C
Density 2 To 6 g/cm3 at 20°C
Quantity 20 kg
MDL Number MFCD00011232
Synonym Silicic acid, Silicic acid
Molecular Weight (g/mol) 60.08 (Anhydrous Basis)
Chemical Name or Material Silicic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.