missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, Supelco™

Product Code. 32238556
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32238556 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32238556 Supplier Supelco Supplier No. 1019051000
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Granules, desiccant ∼ 0.2 to 1 mm

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silica gel
Boiling Point 2230°C
Melting Point 16°C to 19°C (lit.)
CAS 112926-00-8
Quantity 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.