Learn More
Macherey-Nagel™ Silica 60 Adsorbent
Product Code. 10296471 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
1 kg
Particle Size:
0.025 to 0.04 mm
63 μm
Unit Size:
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Particle Size unitSize
10296471 1 kg 0.025 to 0.04 mm 1kg
10023091 1 kg 63 μm 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10296471 Supplier Macherey-Nagel™ Supplier No. 815300.1
 Restricted Item
Estimated Shipment: 12-08-2026
Specifications
Particle Size 0.025 to 0.04 mm
Pore Size ∼60 Å
Technique Column Chromatography
Quantity 1 kg
Surface Area (Metric) ∼500 m2/g
Type Silica 60
For Use With (Application) Low Pressure Column Chromatography