Learn More
Sildenafil citrate, MedChemExpress
Product Code. 16481156
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481156 200 mg 200mg
16436626 100 mg 100mg
16432417 50 mg 50mg
16421756 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16481156 Supplier Medchem Express Supplier No. HY15025A
 Restricted Item
Estimated Shipment: 30-11-2026
MedChemExpress Sildenafil citrate is a potent phosphodiesterase type 5 (PDE5) inhibitor with IC50 of 5.22 nM.
Sildenafil citrate is a potent phosphodiesterase type 5 (PDE5) inhibitor with IC50 of 5.22 nM.
Specifications
Chemical Name or Material Sildenafil citrate
CAS 171599-83-0
Color White
Purity Grade Notes Research
Molecular Formula C28H38N6O11S
Quantity 200 mg
Synonym UK-92480 citrate
Solubility Information DMSO : 50 mg/mL (75.00 mM; Need ultrasonic) ∣H2O : 2 mg/mL (3.00 mM; Need ultrasonic)
SMILES O=C1C(N(C)N=C2CCC)=C2N=C(C3=CC(S(=O)(N4CCN(C)CC4)=O)=CC=C3OCC)N1.O=C(CC(C(O)=O)(O)CC(O)=O)O
Molecular Weight (g/mol) 666.7
Formula Weight 666.7
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation