Learn More
Brand™ Silberbrand™ Eterna™ AR-GLAS™ Class B Measuring Pipets
Product Code. 11893970 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
20 mL
25 mL
Graduations:
0.01 mL
0.02 mL
0.05 mL
0.1 mL
Tolerance:
±0.008 mL
±0.010 mL
±0.015 mL
±0.05 mL
±0.08 mL
±0.15 mL
Color:
2 x Green
Black
Blue
Green
Orange
Red
White
Yellow
Unit Size:
Pack of 12
Pack of 6
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Graduations Tolerance Color Unit Size
11893970 25 mL 0.1 mL ±0.15 mL White Pack of 6
11778158 2 mL 0.01 mL ±0.015 mL White Pack of 12
12055302 5 mL 0.01 mL ±0.008 mL 2 x Green Pack of 12
11853970 2 mL 0.02 mL ±0.015 mL Black Pack of 12
12065302 1 mL 0.01 mL ±0.010 mL Yellow Pack of 12
12075302 1 mL 0.1 mL ±0.010 mL Red Pack of 12
11873970 5 mL 0.1 mL ±0.05 mL Blue Pack of 12
11863970 5 mL 0.05 mL ±0.05 mL Red Pack of 12
11883970 20 mL 0.1 mL ±0.15 mL Green Pack of 6
11788158 10 mL 0.1 mL ±0.08 mL Orange Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11893970 Supplier Brand™ Supplier No. 27079
  • ONLINE EXCLUSIVE PRICE
    61.20 EUR
    40.34 EUR / Pack of 6
    Save 20.86 EUR
    34% Off
Estimated Shipment: 14-12-2026
Manufactured from AR-GLAS™ soda lime glass, this Class B Measuring Pipet is ideal in most research, scientific, and other laboratory environments.
  • Manufactured from AR-GLAS™ Soda lime glass.
  • Type 3, with zero value at the top.
  • Amber stain marking.
  • Class B.
  • Compliant with DIN EN ISO 835 standards.
  • Calibrated to deliver (TD, Ex).
TRUSTED_SUSTAINABILITY
Specifications
Graduations 0.1 mL
Tolerance ±0.15 mL
Color White
Volume (Metric) 25 mL