Learn More
SymLine™ SignalBox
Product Code. 16583532
Change view
Click to view available options
Design:
1-channel
2-channel
5-channel
Plug Type:
EU, UK, US
EU-style
UK-style
US-style
Unit Size:
1 sheet
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Design Plug Type Unit Size
16583532 2-channel EU-style Each
17307419 2-channel UK-style Each
17317419 2-channel US-style Each
18686468 1-channel EU, UK, US 1 sheet
18666498 5-channel EU, UK, US 1 sheet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16583532 Supplier SymLine™ Supplier No. 106548
  • ONLINE EXCLUSIVE PRICE
    552.00 EUR
    456.75 EUR / Pack of 1
    Save 95.25 EUR
    17% Off
Estimated Shipment: 01-12-2026
This built-in signal box monitors up to five filling levels at once, making it ideal for integration into lab furniture
  • Clever way to change a disposal container without interrupting the inflow.
  • LEDs and acoustic buzzer warn you in due time before the filling level of your collection containers reaches a critical height.
  • When container is changed, the alert can be muted via the new touchpad.
  • Includes touchless alarm-resetting fields for up to five containers, up to five switch inputs and up to five switch outputs for peripheral units, and power supply cable with open end and clamp.
Specifications
Design 2-channel
Type Liquid Spill Alarm
For Use With (Equipment) Safety Waste Cap and Electronic Level Controllers
Includes SignalBox2, Power Supply Cable with Open End and Clamp
Dimensions (L x W x H) 36 x 139 x 74 mm
Weight (English) 1.32 lb.
Weight (Metric) 0.6 kg
Plug Type EU-style