Learn More
Scotch-Brite™ Sienna Diamond Floor Pads
Product Code. 17128860
Change view
Click to view available options
Diameter (Metric):
406 mm
432 mm
505 mm
Unit Size:
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
17128860 406 mm Pack of 5
17148860 432 mm Pack of 5
17138860 505 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17128860 Supplier Scotch-Brite™ Supplier No. 7100200175
  • ONLINE EXCLUSIVE PRICE
    353.00 EUR
    212.67 EUR / Pack of 5
    Save 140.33 EUR
    39% Off
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
For cleaning and conditioning worn or dull marble, terrazzo, slate, and polished concrete flooring. When floor appearance reaches a medium shine, the Scotch Brite™ Purple Diamond Floor Pad Plus can be used to further enhance the floor's appearance.
  • Simultaneously cleans and polishes flooring to a brilliant gloss finish without the use of chemicals
  • Ideal for worn or dull marble, terrazzo, vinyl, slate, and polished concrete floors
  • Easy to use with no expensive or harmful chemicals needed, only water
  • Recommended for use with low and standard speed machines
TRUSTED_SUSTAINABILITY
Specifications
Product Type Floor Pad
Color Brown
Diameter (Metric) 406 mm
For Use With (Application) Burnishing
For Use With (Equipment) Low and Standard Speed Machines
Style Round