Learn More
Bohlender™ SICCO™ Glove Box Pure with Transfer Chamber
Product Code. 17107684
Change view
Click to view available options
Material:
Acrylic Glass, Polyethylene
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
17107684 Acrylic Glass, Polyethylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17107684 Supplier Bohlender™ Supplier No. V 174008
  • ONLINE EXCLUSIVE PRICE
    10136.94 EUR
    7621.83 EUR / Pack of 1
    Save 2515.11 EUR
    24% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Glove Box Pure with Transfer Chamber Aluminum frame with two glove ports and one pair of gloves.
  • Aluminum frame with panels made of acrylic glass
  • Base panel made of polyethylene
  • Front panel with two glove ports and one pair of gloves size 9.75 made of UV-resistant EPDM
  • Transfer chamber with two doors on the right, external door with latch to prevent accidental opening
  • Tray extends into workspace with opening of the internal door for easy removal of inserted products
  • Integrated ultraviolet lamp with life-time monitoring on the top side for air and surface disinfection in the workspace
  • Exchangeable lamp
  • Ultraviolet lamp is an ozone-free radiation source that does not emit any radiation below 220 nm
  • Robust and light-weight aluminum construction
  • Superb all-round visibility
  • Non-slip rubber feet
  • Cable lead-ins for electric devices
  • Large front opening
Specifications
Material Acrylic Glass, Polyethylene
Depth (English) 24 in.
Depth (Metric) 600 mm
Height (English) 28 in.
Height (Metric) 700 mm
Voltage 230 V
Volume (Metric) 290 L
Weight (English) 48.5 lb.
Weight (Metric) 22 kg
Width (English) 48 in.
Width (Metric) 1210 mm
Frequency 50 Hz