Learn More
Hanna Instruments™ Shockproof Silicone Boot
Product Code. 11534457
Change view
Click to view available options
Color:
Blue
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11534457 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11534457 Supplier Hanna Instruments™ Supplier No. HI710015
  • ONLINE EXCLUSIVE PRICE
    149.85 EUR
    59.72 EUR / Pack of 1
    Save 90.13 EUR
    60% Off
Estimated Shipment: 03-12-2026
Specifications
Product Type Shockproof Silicone Boot
Color Blue
Material Silicone
For Use With (Equipment) Designed to protect your instrument and prevent damage due to accidental impact