Learn More
Fisherbrand™ Shell Vial Kit, Clear Glass, PE Plug
Product Code. 11516074 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
2.3 mL
5.5 mL
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11516074 5.5 mL Pack of 100
11555914 2.3 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11516074
Only null left
Add to basket
Add to basket
Recommended for HPLC usage.
  • Shell vials and the appropriate plugs can also be supplied as a 2in1 KIT
  • Handy and inexpensive vial/ closure combination for uncritical analyses
TRUSTED_SUSTAINABILITY
Specifications
Description Shell Vial
Color Clear
Diameter (Metric) Outer 14.65 mm
Material Glass
Volume (Metric) 5.5 mL
No. per Pack 100
Closure Size 15
Marking Spot No
Height (Metric) 44.6 mm