Learn More
Asecos™ Shelf Guard Rail System
Product Code. 15836501 Shop All asecos Products
Change view
Click to view available options
For Use With (Equipment):
V90 models
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15836501 V90 models Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15836501 Supplier Asecos™ Supplier No. 35099
  • ONLINE EXCLUSIVE PRICE
    89.25 EUR
    76.00 EUR / Pack of 1
    Save 13.25 EUR
    14% Off
 Restricted Item
Estimated Shipment: 01-03-2027
Secures items safely on shelfs in safety storage cabinets.
  • Accessory item
  • Manufactured from powder-coated sheet steel
  • For V90 cabinets
Specifications
Dimensions (D x W x H) 40 x 719 x 80 mm
For Use With (Application) Securing items on shelves
For Use With (Equipment) V90 models
Weight (Metric) 0.75 kg
Material Steel
Product Type Guard rail system