Learn More
BOLA Sheets
Product Code. 10783491
Change view
Click to view available options
Width (Metric):
150 mm
300 mm
600 mm
Thickness:
0.025 mm
0.05 mm
0.25 mm
0.5 mm
0.75 mm
1 mm
1.5 mm
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Thickness unitSize
10783491 300 mm 0.25 mm Each
10470342 300 mm 0.75 mm Each
10689981 300 mm 1.5 mm Each
10064800 600 mm 0.5 mm Each
10655601 600 mm 0.75 mm Each
10154731 600 mm 1 mm Each
10054860 150 mm 0.025 mm Each
10003531 150 mm 0.05 mm Each
10461371 300 mm 0.025 mm Each
10228590 300 mm 0.05 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10783491 Supplier BOLA Supplier No. S183346
Only null left
Add to basket
Add to basket
Sheets, Non-absorbant
Delivered in rolls with a length of 1000 mm.
Specifications
Absorbancy Non-absorbent
Width (Metric) 300 mm
Thickness 0.25 mm