Learn More
Karl Hecht™ Sharps Disposal Box
Product Code. 10421522
Change view
Click to view available options
Capacity (Metric):
1.8 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10421522 1.8 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10421522 Supplier Karl Hecht™ Supplier No. 49070002
Estimated Shipment: 09-12-2026
For the safe disposal of used needles and clinical sharps, according to UN regulation.
  • Made of polypropylene
  • UN examination includes emission test, GS test, license for the transport of infectious waste, filling mark and cover with incorporated closure
  • Yellow and red in color
Specifications
Capacity (Metric) 1.8 L
Color Yellow
For Use With (Application) Safe disposal of used needles and clinical sharps, according to UN regulation