Learn More
Epredia™ Shandon™ Suresorb™ Clinical Pads and Formaldehyde Neutralizer Pads
Product Code. 12552736 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Formaldehyde Neutralizer Pad
Quantity:
100/Cs.
400/Cs.
50/Cs.
Unit Size:
Pack of 100
Pack of 400
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity Unit Size
12552736 Formaldehyde Neutralizer Pad 400/Cs. Pack of 400
12532736 Formaldehyde Neutralizer Pad 50/Cs. Pack of 50
12542736 Formaldehyde Neutralizer Pad 100/Cs. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12552736 Supplier Epredia™ Supplier No. 5641
  • ONLINE EXCLUSIVE PRICE
    1655.00 EUR
    1600.21 EUR / Pack of 400
    Save 54.79 EUR
    3% Off
 Restricted Item
Estimated Shipment: 11-03-2027
Absorb and lock in liquids with Epredia™ Shandon™ Suresorb™ Clinical Pads and Formaldehyde Neutralizer Pads.
  • Fiber resists tearing, even when saturated
  • Formaldehyde neutralizer pads reduce exposure by chemically bonding with formaldehyde, resulting in the encapsulation of vapors—ideal for grossing areas and autopsy work
  • Clinical pads work as surgical cutting board pads, dissecting pads, autopsy table pads and liners for body trays and cadaver bags
TRUSTED_SUSTAINABILITY
Specifications
Product Type Formaldehyde Neutralizer Pad
Filter Type Formaldehyde
Length (English) 10 in.
Quantity 400/Cs.
Width (English) 8 in.
Width (Metric) 203.2 mm