Learn More
Epredia™ Shandon™ Cartilage Curved Thumb Forceps, Curved, Fine Point, standard, 5 in. (12.7cm)
Product Code. 15307805 Shop All Epredia Products
Change view
Click to view available options
Length (English):
5 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
15307805 5 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15307805 Supplier Epredia™ Supplier No. 1631
  • ONLINE EXCLUSIVE PRICE
    30.65 EUR
    25.89 EUR / Pack of 1
    Save 4.76 EUR
    15% Off
Estimated Shipment: 18-12-2026
Choose 5 in. (12.7cm) cartilage thumb forceps with curved arms and fine points for quality and precision.
Features:
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards
TRUSTED_SUSTAINABILITY
Specifications
Length (English) 5 in.