Learn More
Scientific Industries SI™ Platform for Vortex Shaker
Accepts several types of microplate and microtube foam inserts. Scientific Industries SI™ Platform for Vortex Shaker is also part of the Multiple Sample Starter Set for Vortex-Genie 2 family of mixers.
Brand:  Scientific Industries SI™ 146-6005-00
Additional Details : Weight : 0.75000kg
Product Code. 15577315
  • 48.70 EUR / Each
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
Description
Description
  • For use with the Vortex-Genie 2 family of mixers, Vortex-Genie Pulse
  • Used in conjunction with Microtube and Microplate Foam Inserts, and Two-Tier Microplate Foam Insert
  • Included in the Multiple Sample Starter Set
Specifications
Specifications
Vortex-Genie 2 family of mixers