missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SGLT3/SLC5A4 Polyclonal specifically detects SGLT3/SLC5A4 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | SGLT3/SLC5A4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | DJ90G24.4, low affinity sodium-glucose cotransporter, Na(+)/glucose cotransporter 3, SAAT1low affinity sodium glucose cotransporter, SGLT2, SGLT3, sodium transporter, Sodium/glucose cotransporter 3, solute carrier family 5 (low affinity glucose cotransporter), member 4, solute carrier family 5 (neutral amino acid transporters, system A), member 4, Solute carrier family 5 member 4 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human SGLT3/SLC5A4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?