Learn More
SGE™ Split/Splitless Liner - Recessed Gooseneck with Wool
Description
- All liners are deactivated using high temperature (420°C) gas phase deactivation
- Supplied in blister packs to prevent contamination
- Reduced active sites mean no unwanted reaction in samples
Specifications
Specifications
| Includes | Recessed gooseneck |
| Quantity | 5/Pk. |
| Length (Metric) | 7.85 cm |
| Packing Material | Quartz Wool |
| Product Type | GC Split/Splitless Liner |
| Diameter (Metric) Inner | 0.4 cm |
| Diameter (Metric) Outer | 0.63 cm |
| For Use With (Equipment) | Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177 |