Learn More
Trajan™ Replacement Needles for Luer-Lock Syringes
Product Code. 11929437
Change view
Click to view available options
Needle Gauge:
14 G
16 G
18 G
19 G
23 G
Length (English) Needle:
2 in.
2.7 in.
4.52 in.
Unit Size:
Pack of 5
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Length (English) Needle Unit Size
11929437 23 G 4.52 in. Pack of 5
11989427 23 G 2.7 in. Pack of 5
11961144 19 G 2 in. Pack of 5
10297312 14 G 4.52 in. Pack of 5
11919437 19 G 4.52 in. Pack of 5
11959427 16 G 2.7 in. Pack of 5
11929427 14 G 2 in. Pack of 5
11971144 18 G 2 in. Pack of 5
11951144 23 G 2 in. Pack of 5
11909437 16 G 4.52 in. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11929437 Supplier Trajan™ Supplier No. 039811
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
Estimated Shipment: 29-10-2026
Bevel tip style. Trajan Scientific and Medical™ Replacement Needles for Luer-Lock Syringes are supplied in packs of 5.
Designed for optimum septum penetration and prevention of septum coring.
  • Standard general purpose needle
  • Made of stainless steel
  • Preferred for manual injection where piercing the septum in exactly the same place is difficult
Specifications
Material Stainless Steel
Needle Gauge 23 G
Diameter (English) Inner 0.012 in.
Diameter (English) Outer 0.024 in.
Diameter (Metric) Inner 0.32 mm
Diameter (Metric) Outer 0.63 mm
For Use With (Application) Gas chromatography
Length (English) Needle 4.52 in.
Product Type Replacement Needle
Product Line Trajan
Length (Metric) Needle 115 mm
Needle Point Style Bevel
Connection Type Luer Lock