Learn More
Trajan™ Autosampler Syringes
Product Code. 10214942
Change view
Click to view available options
Volume (Metric):
0.5 μL
1 μL
5 μL
10 μL
Needle Gauge:
23 G
25 G
26 G
Connection Type:
Fixed
Plunger-In, Repeating Adapter
Removable
For Use With (Equipment):
Carlo Erba/Fisons/Leap/CTC AS200 and AS800, GC autosampler syringe
Carlo Erba/Fisons/Leap/CTC ASD200 and AS80, triPlus
PerkinElmer Autosystem
SGE
Shimadzu AOC 14, 17, 20 and 20i GCs
Varian 8035, 8100 and 8200 GC Autosamplers
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Needle Gauge Connection Type For Use With (Equipment) unitSize
10214942 1 μL 23 G Plunger-In, Repeating Adapter SGE Each
10452561 0.5 μL 23 G Removable Shimadzu AOC 14, 17, 20 and 20i GCs Each
10611813 5 μL 26 G Fixed PerkinElmer Autosystem Each
10143402 5 μL 23 G Fixed PerkinElmer Autosystem Each
10033662 5 μL 26 G Fixed Carlo Erba/Fisons/Leap/CTC AS200 and AS800, GC autosampler syringe Each
10245799 5 μL 23 G Fixed Shimadzu AOC 14, 17, 20 and 20i GCs Each
10577922 10 μL 26 G Removable Shimadzu AOC 14, 17, 20 and 20i GCs Each
10380611 10 μL 23 G Removable Shimadzu AOC 14, 17, 20 and 20i GCs Each
10316772 10 μL 25 G Removable Varian 8035, 8100 and 8200 GC Autosamplers Each
10431031 10 μL 26 G Fixed Carlo Erba/Fisons/Leap/CTC ASD200 and AS80, triPlus Each
15208473 10 μL 26 G Removable Carlo Erba/Fisons/Leap/CTC ASD200 and AS80, triPlus Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 10214942 Supplier Trajan™ Supplier No. 000553
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
Replace syringes for Agilent™ Technologies, Varian™, Perkin Elmer™, Shimadzu™, Carlo Erba™, Fisons™, CTC/Leap™, Hitachi™, and more
Syringe needles are cone point unless otherwise noted.
Order Info
Additional syringes are available for other autosamplers made by the manufacturers represented here, as well as syringes for Unicam™, Waters™, and Siemens™ instruments. For information, contact your Fisher Scientific Customer Service Representative.
Specifications
Material Borosilicate Glass (Barrel), PTFE (Plunger)
Needle Gauge 23 G
Diameter (Metric) Inner Needle 0.15 mm
Diameter (English) Outer Needle 0.02 in.
Diameter (English) Outer 0.31 in.
Diameter (Metric) Outer 8 mm
Diameter (Metric) Outer Needle 0.63 mm
Length (English) Needle 2.7 in.
Product Type Autosampler Syringe
Model 1BR-7-RAX
Quantity Ea.
Volume (Metric) 1 μL
Needle Point Style Cone
For Use With (Equipment) SGE
Connection Type Plunger-In, Repeating Adapter