missing translation for 'onlineSavingsMsg'
Learn More

SGC-iMLLT, MedChemExpress

Product Code. 16480997
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480997 50 mg 50mg
16474708 10 mg 10mg
16455536 100 mg 100mg
16417427 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16480997 Supplier Medchem Express Supplier No. HY112804
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SGC-iMLLT is a first-in-class chemical probe and a potent, selective inhibitor of MLLT1/3-histone interactions with an IC50 of 0.26 μM. SGC-iMLLT shows high binding activity towards MLLT1 YEATS domain (YD) and MLLT3 YD (AF9/YEATS3) with Kds of 0.129 and 0.077 μM, respectively.

SGC-iMLLT is a first-in-class chemical probe and a potent, selective inhibitor of MLLT1/3-histone interactions with an IC50 of 0.26 μM. SGC-iMLLT shows high binding activity towards MLLT1 YEATS domain (YD) and MLLT3 YD (AF9/YEATS3) with Kds of 0.129 and 0.077 μM, respectively.

Specifications

Chemical Name or Material SGC-iMLLT
Physical Form Powder
CAS 2255338-25-9
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H24N6O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 83.33 mg/mL (214.51 mM; Need ultrasonic)
SMILES CN1N=CC2=CC(C(NC3=CC=C(NC(CN4[C@@H](C)CCC4)=N5)C5=C3)=O)=CC=C21
Molecular Weight (g/mol) 388.47
Formula Weight 388.47
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.