missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SFRS14 Polyclonal antibody specifically detects SFRS14 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | SFRS14 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:200 - 1:2000, ELISA |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | Arginine/serine-rich-splicing factor 14, DKFZp686M2322, DKFZp779L2418, KIAA0365arginine/serine-rich 14 splicing factor, putative splicing factor, arginine/serine-rich 14, SFRS14, Splicing factor, arginine/serine-rich 14FLJ21855, SRFS14, SURP and G patch domain containing 2, SURP and G-patch domain-containing protein 2 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 500-700 of human SFRS14 (NP_055699.2).,, Sequence:, EAVGLQDIAPSPAAFPNFEDSTLFGREYIDHLKAWLVSSGCPLQVKKAEPEPMREEEKMIPPTKPEIQAKAPSSLSDAVPQRADHRVVGTIDQLVKRVIEGSLSPKERTLLKEDPAYWFLSDENSLEYKYYKLKLAEMQRMSENLRGADQKPTSADCAVRAMLYSRAVRNLKKKLLPWQRRGLLRAQGLRGWKARRATTGT |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?