Learn More
Cytiva SFM4Transfx-293 Media
Product Code. 11536341
Change view
Click to view available options
Quantity:
500 mL
1000 mL
6 x 1000 mL
Unit Size:
1000mL
500mL
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11536341 1000 mL 1000mL
10257653 6 x 1000 mL Pack of 6
10649783 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11536341 Supplier Cytiva Supplier No. SH30860.02
  • ONLINE EXCLUSIVE PRICE
    91.20 EUR
    67.98 EUR / 1000mL
    Save 23.22 EUR
    25% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Promote transfection using lipofection and support growth of HEK 293 cultures with HyClone™ SFM4Transfx-293 Media, a serum-free, animal-derived component free media.
  • ADCF
  • Supports high transfection efficiency
  • Metabolically designed for high cell yield and recombinant protein/vector production
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
TRUSTED_SUSTAINABILITY
Specifications
Form Liquid
Product Type SFM4Transfx-293
Sterilization Method 0.2 μm Sterile-filtered
With Additives Poloxamer 188, Sodium Bicarbonate
Without Additives No Protein, No Phenol Red, No HEPES, No Glutamine
Quantity 1000 mL
Certifications/Compliance cGMP
Content And Storage 2° to 8°C, away from light