Learn More
Sethoxydim, Dr. Ehrenstorfer
Product Code. 30322313
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322313 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30322313 Supplier Dr. Ehrenstorfer Supplier No. DREC16940000
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sethoxydim
CAS 74051-80-2
Analyte or Component Names Sethoxydim
Shipping Condition Room Temperature
Molecular Formula C17 H29 N O3 S
Certifications/Compliance ISO/IEC 17025
Synonym 2-[1-(Ethoxyimino)butyl]-5-[2-(ethylthio)propyl]-3-hydroxy-2-cyclohexen-1-one, ARD 34/02, Alloxol S, BAS 9052, BAS 90520H, BASF 9052, Cyethoxydim, Expand, Expand Plus, Fervinal, Fervinal plus, Grasidim, NP 55, Nabu, Nabu S, Poast, Poast Plus, Segment, Sertin, Sethoxydim
InChI Formula InChI=1S/C17H29NO3S/c1-5-8-14(18-21-6-2)17-15(19)10-13(11-16(17)20)9-12(4)22-7-3/h12-13,19H,5-11H2,1-4H3
Solution Type Neat
SMILES CCCC(=NOCC)C1=C(O)CC(CC(C)SCC)CC1=O
IUPAC Name 2-(N-ethoxy-C-propylcarbonimidoyl)-5-(2-ethylsulfanylpropyl)-3-hydroxycyclohex-2-en-1-one
Industry Type Environmental Testing
Molecular Weight (g/mol) 327.48
Quantity 10 mg
Formula Weight 327.1868 g/mol