Learn More
Interscience™ Set of 6.4 mm Tubings
Product Code. 16332236
Change view
Click to view available options
For Use With (Equipment):
DiluFlow, DiluFlow Pro, DiluFlow Elite
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
16332236 DiluFlow, DiluFlow Pro, DiluFlow Elite Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16332236 Supplier Interscience™ Supplier No. 513044
Set of 5 dispensing tubings with a 6.4 mm inner diameter.
  • Contains silicone tubings, Tygon™ tubings, and straight connectors
Specifications
Product Type Dispensing Tubing
Autoclavable Autoclavable
For Use With (Application) Adding Dispensing Tubings to Dilutors
For Use With (Equipment) DiluFlow, DiluFlow Pro, DiluFlow Elite
Quantity 5 Tubings/Set