Learn More
Grant Instruments™ Liquid McFarland Standards Set
Product Code. 13160623
Change view
Click to view available options
For Use With (Equipment):
DEN-1 & DEN-1B Densitometer
Unit Size:
5 sets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
13160623 DEN-1 & DEN-1B Densitometer 5 sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13160623 Supplier Grant Instruments™ Supplier No. DEN MCF STDS
  • ONLINE EXCLUSIVE PRICE
    417.00 EUR
    388.48 EUR / 5 sets
    Save 28.52 EUR
    6% Off
 Restricted Item
Estimated Shipment: 22-12-2026
For use with DEN-1 and DEN-1B Densitometers. Grant Instruments™ Liquid McFarland Standards Set is supplied in 16mm dia. glass tubes.
  • Supplied in 16mm ø glass tubes.
  • 0.5 / 1.0 / 2.0 / 3.0 / 4.0 can be decanted into an alternative tube (shelf-life no longer valid).
  • Requires D16 adaptor, supplied with DEN-1 / DEN-1B
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) DEN-1 & DEN-1B Densitometer