Learn More
Thermo Scientific™ Service Deionization Accessories
Product Code. 10314830 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Connection Hose Kit
Wall Holder
For Use With (Equipment):
DI 750 Pressureless Replacement
SDI System
Unit Size:
Each
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) unitSize
10314830 Wall Holder SDI System Each
12419661 Connection Hose Kit SDI System Pack of 2
12674515 Wall Holder DI 750 Pressureless Replacement Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10314830 Supplier Thermo Scientific™ Supplier No. 50155461
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
Use these accessories to optimize Thermo Scientific™ SDI systems.
Use Thermo Scientific™ Service Deionization Accessories to optimize SDI systems.
TRUSTED_SUSTAINABILITY
Specifications
Description Wall holder to fit DI stainless steel cartridges to the wall
For Use With (Equipment) SDI System
Type Wall Holder