Learn More
TPP Techno Plastic Products™ Serological Pipets
Product Code. 11341784
Change view
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
25 mL
50 mL
Graduations:
1/10
1/100
2/10
Color:
Blue
Green
Orange
Purple
Red
Unit Size:
Each
Pack of 50
Pack of 60
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Graduations Color Unit Size
11341784 25 mL 2/10 Red Each
11321784 10 mL 1/10 Orange Each
11391774 2 mL 1/100 Green Each
11311784 5 mL 1/10 Blue Each
11361784 25 mL 2/10 Red Pack of 60
11371784 50 mL 1/10 Purple Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11341784 Supplier TPP Techno Plastic Products™ Supplier No. 94525
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured with the highest grade polystyrene for robustness and extreme break resistance. TPP Techno Plastic Products™ Serological Pipets offers large capacity volumes in a compact, ergonomic design.
  • Accurate volume; highly visible, bidirectional (ascending, descending); over-volume graduations
  • Smooth interiors, prevent cell hang-up or clumping inside the pipette
  • Excellent optical transparency
  • Color coded cotton plugs and dispenser boxes for easy identification
  • Convenient, stackable TPP dispenser box
  • Single wrapped in lint-free paper/plastic blister
  • Slender and short form enables ergonomic working conditions in the sterile hood
  • Small outside diameter of the tube at the tip results in an excellent accessibility of the large pipettes even in small tissue culture flasks
  • Highly drop free pipet tip design
  • Optimized form of the mouthpiece preserves the rubber of the pipet aid
  • Models available with reservoir; reservoir augments the total volume capacity without adding proportionate length to the pipet
Spécification
Material Polystyrene
Graduated Yes
Graduations 2/10
Color Red
Volume (Metric) 25 mL
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.