Description
- Durable ABS construction for scratch resistance and easy cleaning
- Strong magnets on both sides for secure mounting to most metal surfaces
- Rack can stand on flat surfaces without mounting
- Two included accessory attachments keep pipet fillers and bulbs within reach
- Side windows allow for easy viewing of pipet size and inventory
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | Red |
| Holds | Serological Pipets |
| Material | ABS with Mounting Magnets |
| Positions | 4 |
| Shape | Rectangular |
| Stackable | No |
| Sterility | Non-sterile |
| Product Type | Side-mounted Pipet Holder |
| Shelves | 4 Tiers |
| Show More |